Learn More
ACROBiosystems Human Fc gamma RIIB / CD32b Protein, HSA,His Tag
Supplier:  ACROBiosystems CDBH52S950UG
Encompass_Preferred
Recombinant Protein;50UG;Human CD32b Protein, HSA,His Tag (CDB-H52S9) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 217 (Accession # P31994-1).;This protein carries a HSA tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 88.2 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-205-4986