Learn More
ACROBiosystems Human Fc gamma RIIIA / CD16a (V176) Protein, SUMO,His Tag
Catalog No. 502054973
Encompass_Preferred
Recombinant Protein;1MG;Human CD16a (V176), SUMO,His Tag (CDA-H5290) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1 (F176V)).;This protein carries a SUMO tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-205-4973 Supplier ACROBiosystems Supplier No. CDAH52901MG
Recombinant Protein;1MG;Human CD16a (V176), SUMO,His Tag (CDA-H5290) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1 (F176V)).;This protein carries a SUMO tag at the N-terminus and a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.5 kDa. The protein migrates as 45-65 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.