Learn More
ACROBiosystems Human Fc gamma RIIIA / CD16a (V176) Protein, HSA,His Tag
Catalog No. 502054962
Encompass_Preferred
Recombinant Protein;50UG;Human CD16a (V176), HSA,His Tag (CDA-H52S1) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1 (F176V)).;This protein carries a HSA tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 90.6 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.
Catalog No. 50-205-4962 Supplier ACROBiosystems Supplier No. CDAH52S150UG
Recombinant Protein;50UG;Human CD16a (V176), HSA,His Tag (CDA-H52S1) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1 (F176V)).;This protein carries a HSA tag at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 90.6 kDa. The protein migrates as 90-115 kDa under reducing (R) condition (SDS-PAGE) due to Glycosylation.