Learn More
ACROBiosystems Human Fc Gamma RIIIA / CD16a Binding Kit, TR-FRET, Competition, Human, 100 Tests
Catalog No. 5000807424
Encompass_Preferred
The Human Fc gamma RIIIA / CD16a (V176) binding Kit (TR-FRET) is a homogeneous, wash-free assay designed to measure the interaction between human CD16a (V176) and antibody drug candidates or inhibitors. Utilizing time-resolved fluorescence resonance energy transfer technology, this kit facilitates the evaluation of antibody-dependent cellular cytotoxicity functional performance and supports high-throughput screening of inhibitors. It serves as a universal tool to identify the binding ability of antibody drugs to human CD16a (V176).
  • Validated with various antibody subtypes and drugs
  • Simple and fast operation with no washing steps
  • High batch consistency and stable supply
  • Sensitive results with minimal matrix effects
  • High-throughput compatibility
  • Fast completion with results in 1 hr
Catalog No. 50-008-07424 Supplier ACROBiosystems Supplier No. FRT07100TESTS
The Human Fc gamma RIIIA / CD16a (V176) binding Kit (TR-FRET) is a homogeneous, wash-free assay designed to measure the interaction between human CD16a (V176) and antibody drug candidates or inhibitors. Utilizing time-resolved fluorescence resonance energy transfer technology, this kit facilitates the evaluation of antibody-dependent cellular cytotoxicity functional performance and supports high-throughput screening of inhibitors. It serves as a universal tool to identify the binding ability of antibody drugs to human CD16a (V176).
  • Validated with various antibody subtypes and drugs
  • Simple and fast operation with no washing steps
  • High batch consistency and stable supply
  • Sensitive results with minimal matrix effects
  • High-throughput compatibility
  • Fast completion with results in 1 hr