Learn More
Biomatik Corporation Human Fc Region Of Immunoglobulin G, Fcγ Elisa Kit
Supplier:  Biomatik Corporation EKC31222
Encompass_Preferred
Human Fc Region Of Immunoglobulin G, Fcγ Elisa Kit for detecting Fc Region Of Immunoglobulin G, Fc in serum, plasma. Species Reactivity: Human (Homo sapiens). Detection Range: 0.027-20ug/ml. Sensitivity: 0.024ug/mL. Format: 96-Well Plate. Assay Type: Competitive. Assay Time: 1-5h. Target Synonyms: Fc region of immunoglobulin G; Fcγ; Fc region of immunoglobulin G; Fc gamma. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5667