Learn More
Sino Biological Anti-CD89/FCAR Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10414T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD89 (rh CD89; Catalog#10414-H08H; NP_001991.1; Met1-Asn227). CD89 specific IgG was purified by Human CD89 affinity chromatography.
Catalog No. 50-255-5310