Learn More
Sino Biological Recombinant Human CD89/FCAR Protein (ECD, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10414H49HB20
Encompass_Preferred
A DNA sequence encoding the human FCAR (NP_001991.1) (Met1-Asn227) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-240-8566