Learn More
Sino Biological Human FCGR2A Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10374H27H2B100
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Ala36-Met210) of human CD32A (P12318-1) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-1969