Learn More
Sino Biological Recombinant Anti-CD32B/Fcgr2b Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10259R11220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD32b extracellular domain (rh CD32b; Catalog#10259-H08H; Ala 46-Pro 217; NP_001002274.1).
Catalog No. 50-241-1470