Learn More
Sino Biological Recombinant Anti-CD16/FCGR3/Fc gamma RIII Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10389R22120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human CD16a/Fc gamma RIIIa (rh CD16a/Fc gamma RIIIa; Catalog#10389-H08H; P08637-1; Met1-Gln208,176F).
Catalog No. 50-255-5335