Learn More
ACROBiosystems HUMAN FCGR3A 25UG
Catalog No. 501017466
Encompass_Preferred
MABSol Biotinylated Human Fc gamma RIIIA / CD16a (F176) (CDA-H82E8) is expressed from human HEK293 cells. It contains AA Gly 17 - Gln 208 (Accession # P08637-1).
Catalog No. 50-101-7466 Supplier ACROBiosystems Supplier No. CDAH82E825UG
MABSol Biotinylated Human Fc gamma RIIIA / CD16a (F176) (CDA-H82E8) is expressed from human HEK293 cells. It contains AA Gly 17 - Gln 208 (Accession # P08637-1).