Learn More
Raybiotech Inc Human FCGR3A/CD16A ELISA Kit
Supplier:  Raybiotech Inc ELHFCGR3A1
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 1.35 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human FCGR3A.
Catalog No. 50-194-9245