Learn More
Sino Biological Human FCGR3A PAb 20ul (Sino Biological)
Supplier:  Sino Biological 10389T2420
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human CD16a/Fc gamma RIIIa (rh CD16a/Fc gamma RIIIa; Catalog#10389-H08H; P08637-1; Met1-Gln208,176F). CD16a/Fc gamma RIIIa specific IgG was purified by Human CD16a/Fc gamma RIIIa affinity chromatography.
Catalog No. 50-237-3927