Learn More
Sino Biological Recombinant Human CD16b/FCGR3B/Fc gamma RIIIB Protein (NA2 allotype, His & AVI Tag)
Supplier:  Sino Biological 11046H27H1
Encompass_Preferred
A DNA sequence encoding the human FCGR3B (O75015.2) (Met1-Ser200) was expressed with a C-terminal polyhistidine tagged AVI tag at the C-terminus (AVI-his).
Catalog No. 50-255-4951