Learn More
Sino Biological Recombinant Human CD16b/FCGR3B/Fc gamma RIIIB Protein (NA1 allotype, His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11046H27H1B100
Encompass_Preferred
A DNA sequence encoding the human FCGR3B (AAA35881.1) (Met1-Ser200) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-237-2979