Learn More
BIOHIPPO Human FcgRI Microsample
SDP
Catalog No. 5000314627
Encompass
Human FcgRI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgRI in serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 32 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FcgRI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FcgRI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FcgRI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14627 Supplier BIOHIPPO Supplier No. BHE15212132
Human FcgRI Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgRI in serum plasma tissue homogenates and other biological fluids It provides a detection range of 78 13-5000 pg/mL with a sensitivity of 32 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FcgRI Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FcgRI Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FcgRI biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color