Learn More
BIOHIPPO Human FcgRII Microsample
SDP
Catalog No. 5000314626
Encompass
Human FcgRII Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgRII in serum plasma and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 17 39 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FcgRII Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FcgRII Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FcgRII biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14626 Supplier BIOHIPPO Supplier No. BHE15212131
Human FcgRII Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FcgRII in serum plasma and other biological fluids It provides a detection range of 46 88-3000 pg/mL with a sensitivity of 17 39 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FcgRII Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FcgRII Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FcgRII biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color