Learn More
Sino Biological Recombinant Anti-ficolin 1/FCN1 Antibody, Rabbit Monoclonal
Supplier:  Sino Biological 10930R001100
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human FCN1 (rh FCN1; Catalog#10930-H08H; NP_001994.2; Met 1-Ala 326).
Catalog No. 50-255-5061