Learn More
Sino Biological Anti-ficolin 1/FCN1 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10930MM0620
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human FCN1 (rh FCN1; Catalog#10930-H08H; NP_001994.2; Met 1-Ala 326). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-255-5062