Learn More
Sino Biological Anti-ficolin 1/FCN1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10930RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FCN1 (rh FCN1; Catalog#10930-H08H; NP_001994.2; Met 1-Ala 326). FCN1 specific IgG was purified by human FCN1 affinity chromatography.
Catalog No. 50-255-5059