Learn More
Sino Biological Anti-FCRL1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 11536RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human FCRL1 / FCRH1 (rh FCRL1; Catalog#11536-H08H; NP_443170.1; Met1-Asn303). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4699