Learn More
Sino Biological Recombinant Human FCRL5 Protein (ECD, His Tag), HPLC-verified
Catalog No. 502993017
Encompass_Preferred
A DNA sequence encoding the Human FCRL5 (Q96RD9) (Gln16-Gly851) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-299-3017 Supplier Sino Biological Supplier No. 11915H08H100
A DNA sequence encoding the Human FCRL5 (Q96RD9) (Gln16-Gly851) was expressed with a polyhistidine tag at the C-terminus.