Learn More
ACROBiosystems Biotinylated Human FCRL5 Protein, His,Avitag™ (MALS & SPR verified)
Catalog No. 502331753
Encompass_Preferred
Biotinylated Human FCRL5, His,Avitag (FC5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 16 - Gly 851 (Accession # Q96RD9-1).
Catalog No. 50-233-1753 Supplier ACROBiosystems Supplier No. FC5H82E3200UG
Biotinylated Human FCRL5, His,Avitag (FC5-H82E3) is expressed from human 293 cells (HEK293). It contains AA Gln 16 - Gly 851 (Accession # Q96RD9-1).