Learn More
Bon Opus Biosciences Recombinant Human FcRn/B2M Heterodimer 50ug
SDP
Supplier:  Bon Opus Biosciences BP22350UG
Encompass_Preferred
Human FcRn & B2M Heterodimer is produced by our Mammalian expression system and the target gene encoding Ala24-Ser297&Ile21-Met119 is expressed with a 6His tag at the C-terminus. Accession# P55899&P61769. Formulation: Lyophilized from a 0.2 μm filtered solution of PBS, 0.05%Tween 80, pH7.4 Bon Opus Cat# BP223
Catalog No. 50-217-2713