Learn More
ACROBiosystems Human FcRn FCGRT B2M 500ug
Catalog No. 502535079
Encompass_Preferred
Human FCGRTB2M Heterodimer Protein Strep TagTag Free (FCM-H5283) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession # P55899-1 (FCGRT) P61769-1 (B2M)).
Catalog No. 50-253-5079 Supplier ACROBiosystems Supplier No. FCMH5283500UG
Human FCGRTB2M Heterodimer Protein Strep TagTag Free (FCM-H5283) is expressed from human 293 cells (HEK293). It contains AA Ala 24 - Ser 297 (FCGRT) Ile 21 - Met 119 (B2M) (Accession # P55899-1 (FCGRT) P61769-1 (B2M)).