Learn More
Sino Biological Human FEN1 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 100916T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human FEN1, and purified by antigen affinity chromatography.
Catalog No. 50-237-2963