Learn More
Sino Biological Anti-Fetuin B Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 106297T40100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Fetuin B/FETUB, and purified by antigen affinity chromatography.
Catalog No. 50-240-8420