Learn More
Raybiotech Inc Human Fetuin A ELISA Kit
Supplier:  Raybiotech Inc ELHFETUINA1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 0.2 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Fetuin A. Other species not determined.
Catalog No. 50-194-9252