Learn More
Abcam Human Fetuin A ELISA Kit (AHSG), 1 x 96 Tests
Supplier:  Abcam AB1088551X96TESTS
Encompass_Preferred
Human Fetuin A ELISA Kit (AHSG) is a Sandwich (quantitative) ELISA kit for the measurement of Human Fetuin A (AHSG) in Human in Cerebral Spinal Fluid, Saliva, Urine, Plasma, Milk, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2122884