Learn More
Sino Biological Human Fetuin A/FETUA Matched ELISA Antibody Pair Set 5 Plates
Catalog No. 5000786023
Encompass_Preferred
The Human Fetuin-A ELISA Pair Set is for the quantitative determination of Human Fetuin-A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Fetuin-A coated on a 96-well plate. Standards and samples are added to the wells, and any Human Fetuin-A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human Fetuin-A monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Fetuin-A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.
Catalog No. 50-007-86023 Supplier Sino Biological Supplier No. SEKA103185
The Human Fetuin-A ELISA Pair Set is for the quantitative determination of Human Fetuin-A.The Sino Biological ELISA Pair Set is a solid phase sandwich ELISA (Enzyme-Linked Immunosorbent Assay). It utilizes a monoclonal antibody specific for Human Fetuin-A coated on a 96-well plate. Standards and samples are added to the wells, and any Human Fetuin-A present binds to the immobilized antibody. The wells are washed and a horseradish peroxidase conjugated rabbit anti-Human Fetuin-A monoclonal antibody is then added, producing an antibody-antigen-antibody "sandwich". The wells are again washed and TMB substrate solution is loaded, which produces color in proportion to the amount of Human Fetuin-A present in the sample. To end the enzyme reaction, the stop solution is added and absorbances of the microwell are read at 450 nm.