Learn More
Raybiotech Inc Human Fetuin B ELISA Kit
Supplier:  Raybiotech Inc ELHFETUINB1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 4 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects human Fetuin B. No cross-reactivity with rhFetuin A, rhMMP2, rhBMP2, 4, 6, rhCystatin A, B, C, 9, rhTGF beta 1, or rhTGFbeta 2 is observed.
Catalog No. 50-194-9253