Learn More
BIOHIPPO Human FGa Microsample
SDP
Catalog No. 5000314439
Encompass
Human FGa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGa in plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 6 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FGa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FGa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FGa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-14439 Supplier BIOHIPPO Supplier No. BHE15212192
Human FGa Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of FGa in plasma It provides a detection range of 15 63-1000 ng/mL with a sensitivity of 6 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human FGa Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human FGa Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human FGa biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color