Learn More
ACROBiosystems Biotinylated Human FGF-10 / KGF 2 Protein, His,Avitag™ (SPR verified)
Catalog No. 502719641
Encompass_Preferred
Fibroblast Growth Factor 10 (FGF 10) is an evolutionary conserved secreted growth factor mediating mostly mesenchymal to epithelial signaling. FGF 10 belongs to the FGF 7 subfamily and shares similar biochemical and amino acid sequences with its constituent members (FGF3, FGF 7 and FGF 22). As a paracrine FGF, FGF 10 elicits its biological responses by activating the fibroblast growth factor receptor 2b (FGF R 2b), is crucial for governing proximal distal outgrowth as well as patterning and acts upstream of the known apical ectodermal ridge (AER) marker FGF 8. FGF10 is also implicated in pancreatic cancer, and that overexpression of FGFR2b is associated with metastatic invasion.
Catalog No. 50-271-9641 Supplier ACROBiosystems Supplier No. FG0H81Q7200UG
Fibroblast Growth Factor 10 (FGF 10) is an evolutionary conserved secreted growth factor mediating mostly mesenchymal to epithelial signaling. FGF 10 belongs to the FGF 7 subfamily and shares similar biochemical and amino acid sequences with its constituent members (FGF3, FGF 7 and FGF 22). As a paracrine FGF, FGF 10 elicits its biological responses by activating the fibroblast growth factor receptor 2b (FGF R 2b), is crucial for governing proximal distal outgrowth as well as patterning and acts upstream of the known apical ectodermal ridge (AER) marker FGF 8. FGF10 is also implicated in pancreatic cancer, and that overexpression of FGFR2b is associated with metastatic invasion.