Learn More
Raybiotech Inc Human FGF-12 ELISA Kit
Supplier:  Raybiotech Inc ELHFGF121
Encompass_Preferred
For Cell Culture Supernatants, Plasma, and Serum samples. Sensitivity: 32 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes human FGF-12. Other species not yet determined.
Catalog No. 50-194-9254