Learn More
ACROBiosystems Human FGF-17 50ug
Catalog No. 502535067
Encompass_Preferred
Human FGF-17 Protein His Tag premium grade (FG7-H5144) is expressed from E. coli cells. It contains AA Thr 23 - Thr 216 (Accession # O60258-1).
Catalog No. 50-253-5067 Supplier ACROBiosystems Supplier No. FG7H514450UG
Human FGF-17 Protein His Tag premium grade (FG7-H5144) is expressed from E. coli cells. It contains AA Thr 23 - Thr 216 (Accession # O60258-1).