Learn More
Raybiotech Inc Human FGF-21 ELISA Kit
Supplier:  Raybiotech Inc ELHFGF211
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 8 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: Cross Reactivity: This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM12, ADAM8, B7-H3, BMPR-IB, Cadherin-13, Cadherin-4, CD155, CD229, CD48, CD58, CD84, CD99, CEACAM-5, CF XIV, Cystatin A, Cystatin B, Cystatin E/M, Desmoglein 2, DR3, ErbB4, ESAM, Galectin-2, Galectin-9, ICOS, JAM-A, JAM-B, Kallikrein 5, Midkine, Pentraxin 3, Pref-1, Siglec-10, SLAM, SP-D, Syndecan-4, Testican 2, TIM-3, TLR4, TRAIL, ULBP-1.
Catalog No. 50-194-9259