Learn More
Invitrogen™ Human FGF-21 ELISA Kit
Description
Human FGF-21 quantitates human FGF-21 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FGF-21.
Specifications
Specifications
| Accession Number | Q9NSA1 |
| Assay Range | 8-2000 pg/mL |
| Assay Sensitivity | 8 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196FGF; FGF21; FGF-21; FGFL; Fibroblast growth factor; Fibroblast growth factor 21; PRO10196; UNQ3115/PRO10196; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.