Learn More
FUJIFILM BIOSCIENCES INC Human FGF-21, Recombinant Protein, Shenandoah Biotechnology, 20 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10014820UG
Encompass_Preferred
Fibroblast growth factor 21 (FGF-21) is an endocrine hormone that regulates energy homeostasis and exerts cardioprotective functions during heart injury. FGF-21 is expressed in the liver, pancreas, heart, and adipose tissues. FGF-21 signaling is activated through the FGFR1c receptor and B-Klotho co-receptor. FGF-21 is an important regulator of glucose uptake and reduces cell apoptosis under stress conditions.
Catalog No. 50-990-693