Learn More
ACROBiosystems ACROBiosystems Human FGF-21 Protein, His Tag
Supplier:  ACROBiosystems FG1H52431MG
Encompass_Preferred
Recombinant Protein;1MG;Human FGF-21, His Tag (FG1-H5243) is expressed from human 293 cells (HEK293). It contains AA His 29 - Ser 209 (Accession # Q9NSA1-1).;This protein carries a polyhistidine tag at the N-terminus. The protein has a calculated MW of 21.3 kDa. The protein migrates as 25-28 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-210-7367