Learn More
FUJIFILM Biosciences Human FGF-22, Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Catalog No. 50900464
Encompass_Preferred
Fibroblast growth factor 22 (FGF-22) is a mediator of synaptogenesis in the adult nervous system and functions to regulate synapse formation and maturation. FGF-22 is expressed in the inner hair cell and functions to maintain ribbon synapse number to protect functional hearing. In the hippocampus, FGF-22 promotes excitatory synapse formation through binding the FGFR2b and FGFR1b receptors. FGF-22 is also required for axonal circuit remodeling after spinal cord injury.
Catalog No. 50-900-464 Supplier FUJIFILM Biosciences Supplier No. 100681MG
Add to Cart
Add to Cart
Request a Quote
Fibroblast growth factor 22 (FGF-22) is a mediator of synaptogenesis in the adult nervous system and functions to regulate synapse formation and maturation. FGF-22 is expressed in the inner hair cell and functions to maintain ribbon synapse number to protect functional hearing. In the hippocampus, FGF-22 promotes excitatory synapse formation through binding the FGFR2b and FGFR1b receptors. FGF-22 is also required for axonal circuit remodeling after spinal cord injury.