Learn More
ACROBiosystems Biotinylated Human FGF-7, His,Avitag (FG7-H82E7) is expressed from human 293 cells (HEK293). It contains AA Cys 32 - Thr 194 (Accession # P21781-1).
Supplier:  ACROBiosystems FG7H82E725UG
Encompass_Preferred
Fibroblast growth factor (FGF) 7 (is also known as Keratinocyte growth factor (KGF)), a member of FGF family, is initially found to be secreted from mesenchymal cells to repair epithelial tissues. As a well-characterized paracrine growth factor for tissue growth and regeneration, fibroblast growth factor 7 (FGF7) is involved in a number of physiological and pathological processes, including lung disease and cancer. The stromal-derived FGFs, such as FGF7 and FGF10, control epithelial cell resident FGFR2IIIb activities, promote net tissue homeostasis, and restraint tumor cells from progression to malignancy.
Catalog No. 50-223-0616