Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-acidic (FGF-1), Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF10017A250
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF10017A250 250 μg
50-398-424 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AF10017A250 Supplier Gibco™ Supplier No. AF10017A250UG
Add to Cart
Add to Cart

Recombinant Protein

Recombinant Human FGF-acidic is a 16.8 kDa protein consisting of 141 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF1 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein functions as a modifier of endothelial cell migration and proliferation, as well as an angiogenic factor. It acts as a mitogen for a variety of mesoderm- and neuroectoderm-derived cells in vitro, thus is thought to be involved in organogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05230
For Use With (Application) Control, Bioactivity
Format Lyophilized
Formulation Protein with no preservative
Gene ID (Entrez) 2246
Molecular Weight (g/mol) 16.8 kDa
Name Human FGF-acidic (FGF-1), Animal-Free
Purification Method Purified
Quantity 250 μg
Storage Requirements -20°C
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/μg
Gene Alias acidic eye-derived fibroblast growth factor; acidic eye-derived growth factor II; Acidic fibroblast growth factor; aFGF; beta-endothelial cell growth factor; Dffrx; ECGF; ECGFA; ECGFB; ECGF-beta; EDGF; EDGF II; Endothelial cell growth factor; Endothelial cell growth factor alpha; Endothelial cell growth factor beta; endothelial cell growth factor beta and alpha chains; endothelial cell growth factor, alpha; endothelial cell growth factor, beta; endothelial growth factor; Fam; FGF; FGF1; Fgf-1; FGF1 protein; Fgfa; FGF-alpha; Fibroblast growth factor; Fibroblast growth factor 1; fibroblast growth factor 1 (acidic); Fibroblast growth factor 1 (heparin binding); fibroblast growth factor, acidic; GLIO703; HBGF1; HBGF-1; heparin-binding growth factor; Heparin-binding growth factor 1; H-FGF-a; M-FGF-a; Prostatropin; unnamed protein product
Common Name FGF1
Gene Symbol FGF1
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 0.5 ng/ml in the presence of 10 ug/ml heparin, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Sequence MFNLPPGNYK KPKLLYCSNG GHFLRILPDG TVDGTRDRSD QHIQLQLSAE SVGEVYIKST ETGQYLAMDT DGLLYGSQTP NEECLFLERL EENHYNTYIS KKHAEKNWFV GLKKNGSCKR GPRTHYGQKA ILFLPLPVSS D
Content And Storage -20°C
Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 0.5 ng/ml in the presence of 10 ug/ml heparin, corresponding to a specific activity of ≥ 2 x 10^6 units/mg.
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.