Learn More
FUJIFILM Biosciences Human FGF-basic 147 (FGF-2) , Recombinant Protein, Shenandoah Biotechnology, 1 mg vial
Catalog No. 50990736
Encompass_Preferred
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis.  FGF-basic also has cardioprotective functions during heart injury.  FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state.  Degredation of the full length FGF-basic N-terminus results in a truncated FGF-basic 147aa protein, when the protein is isolated from biological sources.  The N-terminus extensions influence the localization of FGF-basic within the cell, but do not affect the biological activity of FGF-basic.  Therefore, there are no detectable differences in biological activity between the full length FGF-basic 154aa and the truncated FGF-basic 147aa recombinant proteins.
Catalog No. 50-990-736 Supplier FUJIFILM Biosciences Supplier No. 100281MG
Basic fibroblast growth factor (FGF-basic), also known as FGF-2, is expressed by endothelial cells and is a mediator of angiogenesis.  FGF-basic also has cardioprotective functions during heart injury.  FGF-basic is a critical component for embryonic stem cell culture systems and is necessary for maintaining cells in an undifferentiated state.  Degredation of the full length FGF-basic N-terminus results in a truncated FGF-basic 147aa protein, when the protein is isolated from biological sources.  The N-terminus extensions influence the localization of FGF-basic within the cell, but do not affect the biological activity of FGF-basic.  Therefore, there are no detectable differences in biological activity between the full length FGF-basic 154aa and the truncated FGF-basic 147aa recombinant proteins.