Learn More
ACROBiosystems Human FGF basic 1mg
Supplier:  ACROBiosystems BFFH51151MG
Encompass_Preferred
Human FGF basic (154aa) Protein premium grade (BFF-H5115) is expressed from E. coli cells. It contains AA Ala 135 - Ser 288 (Accession # NP_001997.5).
Catalog No. 50-253-5469