Learn More
Bon Opus Biosciences Recombinant Human Fibroblast Growth Factor 2/FGF-2/FGFb (Gly132-Ser288) 500ug
SDP
Supplier:  Bon Opus Biosciences C046500UG
Encompass_Preferred
Recombinant Human Fibroblast growth factor 2 is produced by our E.coli expression system and the target gene encoding Gly132-Ser288 is expressed. Accession# P09038. Formulation: Lyophilized from a 0.2 μm filtered solution of 20mM TrisHCl, 150mM NaCl, pH 7.5. Bon Opus Cat# C046
Catalog No. 50-217-2718