Learn More
ACROBiosystems Human FGF basic 50ug
Catalog No. 502535467
Encompass_Preferred
Human FGF basic (154aa) Protein premium grade (BFF-H5115) is expressed from E. coli cells. It contains AA Ala 135 - Ser 288 (Accession # NP_001997.5).
Catalog No. 50-253-5467 Supplier ACROBiosystems Supplier No. BFFH511550UG
Human FGF basic (154aa) Protein premium grade (BFF-H5115) is expressed from E. coli cells. It contains AA Ala 135 - Ser 288 (Accession # NP_001997.5).