Learn More
R&D Systems™ Human FGF basic/FGF2/bFGF Quantikine HS ELISA Kit
Description
Specifications
Specifications
| Assay Range | 0.31 - 20 Not Assigned |
| Assay Sensitivity | 0.07 Not Assigned |
| Conjugate | HRP |
| Cross Reactivity | Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules |
| Product Type | Solid Phase Sandwich ELISA |
| Sample Type | Plasma (Citrate), Plasma (EDTA), Serum, Urine |
| Specificity | Natural and Recombinant Human FGF basic |
| Sufficient For | 1 x 96-well Plate |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.