Learn More
ACROBiosystems Human FGF R2 (IIIb) protein, His Tag (MALS verified)
Catalog No. 502054983
Encompass_Preferred
Recombinant Protein;100UG;Human FGFR2 (IIIb), His Tag (FGB-H5223) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Leu 358 (Accession # P21802-3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.8 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGFR2 (IIIb), His Tag (Cat. No. FGB-H5223) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Tag Free with a linear range of 0.039-0.625 μg/mL (Routinely tested).
Catalog No. 50-205-4983 Supplier ACROBiosystems Supplier No. FGBH5223100UG
Recombinant Protein;100UG;Human FGFR2 (IIIb), His Tag (FGB-H5223) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Leu 358 (Accession # P21802-3).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.8 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGFR2 (IIIb), His Tag (Cat. No. FGB-H5223) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Tag Free with a linear range of 0.039-0.625 μg/mL (Routinely tested).