Learn More
ACROBiosystems Human FGF R2 (IIIc) protein, His Tag (MALS verified)
Catalog No. 502054978
Encompass_Preferred
Recombinant Protein;1MG;Human FGFR2 (IIIc), His Tag (FGC-H5225) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Thr 358 (Accession # P21802-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.9 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGFR2 (IIIc), His Tag (Cat. No. FGC-H5225) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Tag Free with a linear range of 0.04-1.25 μg/mL (Routinely tested).
Catalog No. 50-205-4978 Supplier ACROBiosystems Supplier No. FGCH52251MG
Recombinant Protein;1MG;Human FGFR2 (IIIc), His Tag (FGC-H5225) is expressed from human 293 cells (HEK293). It contains AA Pro 154 - Thr 358 (Accession # P21802-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 24.9 kDa. The protein migrates as 38-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human FGFR2 (IIIc), His Tag (Cat. No. FGC-H5225) at 5 μg/mL (100 μL/well) can bind Biotinylated Human FGF basic, Tag Free with a linear range of 0.04-1.25 μg/mL (Routinely tested).