Learn More
ACROBiosystems ACROBiosystems Human FGF R4 / CD334 Protein, Fc Tag
Supplier:  ACROBiosystems FG4H5253100UG
Encompass_Preferred
Recombinant Protein;100UG;Human FGF R4, Fc Tag (FG4-H5253) is expressed from human 293 cells (HEK293). It contains AA Leu 22 - Asp 369 (Accession # P22455-1).;This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 64.9 kDa.
Catalog No. 50-210-6326